Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methyl-2-pyrrolidone (NMP) ≥99.5%, ALEG®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: N-Methyl-alpha-pyrrolidone1-Methyl-2-pyrrolidoneNMPN-Methyl-2-pyrrolidoneN-Methylpyrrolidone1-Methyl-2-pyrrolidinoneN-MethylpyrrolidinoneN-methylpyrrolidoneN-Methyl-gamma-butyrolactamN-Methyl-2-pyrrolidinoneM-Pyrol

Management of Change (MOC) category = R

BAKR6407-10 BAKR6407-07 6407-10 BAKR6407-58 BAKR6407-QM BAKR6407-54 BAKR6407-59 BAKR6407-QM BAKR6407-59 BAKR6407-54 BAKR6407-58 6407-07
JT6407-7EA 637.22 USD InStock
CAJT6407-59 JT6407-7 CAJT6407-7 JT6407-58 CAJT6407-58 JT6407-59 JT6407-10 JT6407-54 CAJT6407-QM CAJT6407-10 CAJT6407-54 JT6407-QM
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, ALEG®, J.T.Baker®
N-Methyl-2-pyrrolidone (NMP)
Positive photoresist stripper (NMP).


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202 °C (1013 hPa)
Melting Pt: –24 °C
Density: 1.033 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 12,06197
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/54/2298854.jpg
Order Now
Specification Test Results
For Microelectronic Use
Assay (C₅H₉NO) ≥99.5 %
Color (APHA) ≤20
pH of 5% Aqueous Solution, For Information Only
Water (H₂O)(by Karl Fischer titrn) ≤0.05 %
Free Amines (as CH₃NH₂) ≤50 ppm
Chloride (Cl) ≤0.5 ppm
Phosphate (PO₄) ≤1 ppm
Trace Impurities - Aluminum (Al) ≤100.0 ppb
Arsenic and Antimony (as As) ≤100.0 ppb
Trace Impurities - Barium (Ba) ≤100.0 ppb
Trace Impurities - Boron (B) ≤100.0 ppb
Trace Impurities - Calcium (Ca) ≤300 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Cobalt (Co) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤100.0 ppb
Trace Impurities - Iron (Fe) ≤100.0 ppb
Trace Impurities - Lead (Pb) ≤100 ppb
Trace Impurities - Lithium (Li) ≤100.0 ppb
Trace Impurities - Magnesium (Mg) ≤100.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Nickel (Ni) ≤100.0 ppb
Trace Impurities - Potassium (K) ≤100 ppb
Trace Impurities - Silver (Ag) ≤100.0 ppb
Trace Impurities - Strontium (Sr) ≤100.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Tin (Sn) ≤100 ppb
Trace Impurities - Titanium (Ti) ≤100.0 ppb
Trace Impurities - Vanadium (V) ≤100.0 ppb
Trace Impurities - Zinc (Zn) ≤100.0 ppb
Particle Count at point of fill - 0.2 µm and greater (Rion KS42AF)
Particle Count at point of fill - 0.5 µm and greater (Rion KS42AF)
Particle Count at point of fill - 1.0 µm and greater (Rion KS42AF)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty