Would you like to log in again?
J.T.Baker® BAKERBOND®, SPE Columns
Supplier: Avantor
J.T.Baker® BAKERBOND spe™ BAKERBOND®
BAKR7086-07 BAKR7020-07 BAKR7094-03 BAKR7213-07 7020-01 7087-03 BAKR7020-02 BAKR7095-03 7087-06 7217-06 7086-03 BAKR7095-03 BAKR7020-02 7213-27 BAKR7087-06 BAKR7021-07 BAKR7211-03 7086-07 7086-06 7020-07 7214-07 BAKR7420-07 BAKR7094-03 BAKR7087-03 BAKR7214-07 BAKR7213-27 BAKR7090-29 BAKR7086-06 7213-07 7095-03 BAKR7211-03 BAKR7020-07 7020-02 BAKR7086-06 BAKR7020-27 BAKR7090-03 BAKR7090-03 BAKR7091-03 BAKR7090-29 BAKR7189-03 7094-03 BAKR7216-06 BAKR7091-03 BAKR7086-26 BAKR7020-01 7216-06 BAKR7213-07 7420-07 7021-03 BAKR7087-03 BAKR7214-07 BAKR7213-27 BAKR7020-01 BAKR7189-03 BAKR7217-06 BAKR7021-07 BAKR7020-26 7090-29 BAKR7086-07 BAKR7086-03 7189-03 BAKR7021-03 7211-03 BAKR7217-06 BAKR7020-26 BAKR7216-06 7086-26 BAKR7086-03 7090-07 BAKR7420-07 BAKR7020-03 BAKR7213-07 BAKR7020-27 7091-03 BAKR7021-03 BAKR7020-06 BAKR7086-26 BAKR7020-03 BAKR7090-07 BAKR7087-06 7020-06 BAKR7020-06
JT72176CS 2544.02 USD InStock
CAJT70206 CAJT70203 CAJT70202 CAJT70201 CAJT721327 CAJT71893 CAJT72147 JT72176 CAJT7189-3 CAJT74207 CAJT70207 JT70873 JT72137 CAJT7213-27 JT70953 JT70876 CAJT7095-3 CAJT708626 CAJT7090-29 JT70913 JT702026 JT702027 CAJT70217 CAJT7213-7 CAJT70213 CAJT7211-3 JT7213-7 CAJT72113 CAJT7086-26 JT72147 JT70202 JT70201 JT74207 CAJT7020-1 CAJT7087-3 JT70203 JT70206 CAJT7020-2 JT70207 CAJT7020-7 CAJT7020-6 CAJT7087-6 CAJT7217-6 CAJT70943 CAJT70866 CAJT70863 JT7189-03 CAJT709029 JT72113 CAJT70903 CAJT70867 JT70213 10046-880 CAJT702026 JT70217 CAJT702027 CAJT7094-3 CAJT72166 JT721327 CAJT7090-07 JT708626 CAJT70953 CAJT70876 CAJT70873 CAJT7420-7 CAJT72137 CAJT7091-3 CAJT70913 JT72166 CAJT7086-3 JT70863 JT70943 JT70866 CAJT72176 JT70867 CAJT7021-3 JT70903 JT709029 CAJT7086-6 CAJT7216-6 CAJT7086-7 CAJT7214-7
J.T.Baker® BAKERBOND®, SPE Columns
Chromatography Columns
Disposable polypropylene columns are prepacked with selective reversed phase, polar, and ion exchange organosilanes bonded to 40 µm, 60 Å, and 275 Å silica gel absorbents and size exclusion gels.
  • Silica based
  • Processing by vacuum or positive pressure
The highly surface-active sorbents, contained by two 20 µm polyethylene frits, provide high capacity. This configuration allows extraction of compounds from sample solution volumes of a few hundred microliters to several milliliters.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty