Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium bicarbonate

Management of Change (MOC) category = R

J.T. Baker®
BAKR7451-26 BAKR7451-26 BAKR7451-07 BAKR7451-07
JT7451-26EA 2944.82 USD InStock
JT7451-26 CAJT7451-07 JT7451-07 CAJT7451-26
Sodium hydrogen carbonate 99.0-100.5% (dried basis), powder USP, ACS, J.T.Baker®
Sodium hydrogen carbonate
Caution: This product is not intended for use in hemodialysis.


Formula: NaHCO₃
MW: 84.01 g/mol
Density: 2.20 g/cm³
MDL Number: MFCD00003528
CAS Number: 144-55-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/81/16640581.jpg
Order Now
Specification Test Results
Meets U.S.P Requirements
Meets ACS Reagent Chemical Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
ACS - Ammonium (NH₄) ≤5 ppm
ACS - Assay (NaHCO₃) (by acidimetry) 99.7 - 100.3 %
ACS - Calcium (Ca) ≤0.02 %
ACS - Chloride (Cl) ≤0.003 %
ACS - Heavy Metals (as Pb) ≤5 ppm
ACS - Insoluble Matter ≤0.015 %
ACS - Iron (Fe) ≤0.001 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Potassium (K) ≤0.005 %
ACS - Sulfur Compounds (as SO₄) ≤0.003 %
USP - Assay (NaHCO₃) (dried basis) 99.0 - 100.5 %
USP - Chloride (Cl) ≤0.015 %
USP - Identification Passes Test
USP - Insoluble Substances Passes Test
USP - Limit of Ammonia ≤20 ppm
USP - Limit of Sulfur Compounds ≤0.015 %
USP - Loss on Drying ≤0.25 %
USP - Normal Carbonate Passes Test
USP - Arsenic (As) ≤2 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
This product is not intended for use in hemodialysis.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty