Would you like to log in again?
J.T.Baker® BAKERBOND Speedisk® narc-2™, Column, 10 µm
Supplier: Avantor
BAKERBOND® Speedisk™
BAKR8175-04 BAKR8175-04
JT81754EA 422.36 USD InStock
JT81754 CAJT8175-04
J.T.Baker® BAKERBOND Speedisk® narc-2™, Column, 10 µm
Chromatography Columns
Ultra-clean, PP columns with BAKERBOND® HPLC media are engineered to save time, minimize work and final solvent volumes and improve performance. The built-in column guard filter and higher analyte concentration may reduce or eliminate prefiltration and concentration steps.
  • Run times up to nine times faster than conventional SPE columns
  • Unique laminar configuration
  • Silica based
  • Extraction columns for drugs of abuse
General applications: Hydrophobic/basic analytes (cocaine, benzoylecgonine).
These SPE columns can be used in mixed mode.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty