Synonyms:
2-Propanone
, DMK
, Propanone
, Dimethyl ketone
Management of Change (MOC) category = N
BAKR9002-88
BAKR9002-02
BAKR9002-33
BAKR9002-3H
BAKR9002-99
BAKR9002-3H
BAKR9002-03
BAKR9002-03
BAKR9002-98
BAKR9002-99
BAKR9002-7R
BAKR9002-98
BAKR9002-6K
BAKR9002-7R
BAKR9002-50
9002-88
BAKR9002-02
BAKR9002-6K
BAKR9002-YE
BAKR9002-33
BAKR9002-50
BAKR9002-XV
BAKR9002-YE
BAKR9002-XV
JT9002-YECS
3156
USD
InStock
JT9002-YE
JT9002-3
JT9002-2
CAJT9002-YE
JT9002-3H
JT9002-6K
CAJT9002-3H
CAJT9002-6K
CAJT9002-7R
CAJT9002-50
JT9002-7R
JT9002-50
JT9002-88
JT9002-99
JT9002-33
CAJT9002-XV
CAJT9002-98
JT9002-XV
JT9002-98
CAJT9002-2
CAJT9002-3
CAJT9002-33
CAJT9002-99
CAJT9002-88
Acetone ≥99.3% (by GC), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Acetone
- Packaged under nitrogen
- Filtered through a 0.2 µm filter to remove particulate matter