Synonyms:
Dimethyl ketoneDimethylformaldehydeDMKPropanone2-Propanone
BAKR9006-3C
BAKR9006-98
BAKR9006-03
BAKR9006-7R
BAKR9006-98
BAKR9006-01
BAKR9006-R
BAKR9006-3C
BAKR9006-01
BAKR9006-06
BAKR9006-8P
BAKR9006-ZT
BAKR9006-01
BAKR9006-07
BAKR9006-03
BAKR9006-99
BAKR9006-ZX
BAKR9006-9Q
BAKR9006-22
BAKR9006-07
BAKR9006-8P
BAKR9006-99
BAKR9006-05
BAKR9006-ZX
BAKR9006-22
BAKR9006-9Q
BAKR9006-R
BAKR9006-05
BAKR9006-7R
BAKR9006-06
BAKR9006-33
BAKR9006-33
BAKR9006-03
JT9006-3CCS
3247.29
USD
InStock
JT9006-3C
SCJT9006-3
JT9006-9Q
JT9006-R
JT9006-98
CAJT9006-3
JT9006-99
CAJT9006-ZX
JT9006-33
CAJT9006-1
CAJT9006-6
CAJT9006-9Q
CAJT9006-7
CAJT9006-5
CAJT9006-7R
CAJT9006-22
JT9006-1
JT9006-8P
CAJT9006-3C
JT9006-3
JT9006-5
JT9006-7
JT9006-ZX
CAJT9006-R
JT9006-22
CAJT9006-98
JT9006-01
JT9006-ZT
CAJT9006-8P
JT9006-7R
JT9006-06
CAJT9006-99
CAJT9006-33
Acetone ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Acetone
- Produced using proven distillation methods and rigid testing protocols
- Statistical Process Control used in purification and packaging to ensure lot-to-lot consistency
- Statistical Quality Control and suitability-for-use tests to ensure product performance