Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile ≥99.9% (by GC), BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methane, cyano-Ethyl nitrileMethyl cyanideEthanoic acid nitrileMethanecarbonitrileEthanenitrileCyanomethaneACN

Management of Change (MOC) category = N

BAKR9017-76 BAKR9017-3F BAKR9017-3Z BAKR9017-1Z BAKR9017-7M BAKR9017-WK BAKR9017-97 9017-88 BAKR9017-7R BAKR9017-8T BAKR9017-Z1 BAKR9017-02 BAKR9017-98 BAKR9017-N7 BAKR9017-2M BAKR9017-6W 9017-88 BAKR9017-H1 BAKR9017-03 BAKR9017-J9 BAKR9017-U5 BAKR9017-S1 BAKR9017-J3 BAKR9017-48 BAKR9017-3A BAKR9017-YZ BAKR9017-9U BAKR9017-7Q BAKR9017-J3 9017-69 BAKR9017-6K BAKR9017-98 9017-97 BAKR9017-E5 BAKR9017-92 BAKR9017-9P BAKR9017-D5 BAKR9017-8L BAKR9017-6M BAKR9017-WK BAKR9017-50 BAKR9017-8T BAKR9017-YZ BAKR9017-48 BAKR9017-76 BAKR9017-ZS BAKR9017-9C BAKR9017-9U BAKR9017-99 BAKR9017-99 BAKR9017-03 BAKR9017-7Q BAKR9017-3A BAKR9017-D5 BAKR9017-XN BAKR9017-S1 BAKR9017-50 BAKR9017-U5 BAKR9017-02 BAKR9017-6W BAKR9017-E5 BAKR9017-3Z BAKR9017-47 BAKR9017-33 BAKR9017-5J BAKR9017-N7 BAKR9017-6M BAKR9017-5P BAKR9017-XN 9017-69 BAKR9017-7M BAKR9017-92 BAKR9017-6K BAKR9017-5P BAKR9017-9C BAKR9017-8L BAKR9017-47 BAKR9017-3F BAKR9017-YU BAKR9017-Z1 BAKR9017-9P BAKR9017-V1 BAKR9017-H1 BAKR9017-1Z BAKR9017-2M BAKR9017-7R BAKR9017-J9 BAKR9017-ZS BAKR9017-YU 9017-33 BAKR9017-V1 BAKR9017-5J
JT9017-2MCS 5959.83 USD InStock
CAJT9017-7R CAJT9017-7Q JT9017-2M JT9017-33 JT9017-6W JT9017-76 CAJT9017-47 CAJT9017-88 CAJT9017-U5 CAJT9017-3Z JT9017-H1 JT9017-2 JT9017-3 CAJT9017-48 JT9017-D5 JT9017-3A TXCA9017-97 JT9017-3F CAJT9017-E5 CAJT9017-Z1 JT9017-7M JT9017-XN CAJT9017-8L JT9017-9U CAJT9017-2M JT9017-5P CAJT9017-76 JT9017-S1 CAJT9017-6W JT9017-69 CAJT9017-33 JT9017-1Z CAJT9017-2 CAJT9017-3 CAJT9017-H1 CAJT9017-3A JT9017-WK CAJT9017-3F JT9017-6K CAJT9017-D5 CAJT9017-XN CAJT9017-7M JT9017-6M JT9017-8T CAJT9017-5P JT9017-YU CAJT9017-ZS JT9017-50 JT9017-V1 JT9017-99 CAJT9017-S1 JT9017-YZ JT9017-97 JT9017-98 CAJT9017-9U CAJT9017-69 JT9017-N7 JT9017-J3 CAJT9017-1Z JT9017-9C JT9017-J9 JT9017-5J CAJT9017-6M JT9017-ZS CAJT9017-6K CAJT9017-WK JT9017-9P CAJT9017-50 CAJT9017-V1 CAJT9017-92 JT9017-7Q JT9017-7R JT9017-88 CAJT9017-98 JT9017-U5 CAJT9017-8T CAJT9017-YU JT9017-48 CAJT9017-YZ CAJT9017-99 JT9017-47 JT9017-3Z CAJT9017-9C CAJT9017-J3 CAJT9017-N7 CAJT9017-J9 JT9017-E5 JT9017-8L CAJT9017-9P JT9017-92 CAJT9017-5J JT9017-Z1
Acetonitrile ≥99.9% (by GC), BAKER ANALYZED® HPLC, Ultra Gradient for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Acetonitrile


Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/10/718910.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.005
Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 200 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤190
Gradient Elution Test (a.u.) - 254 nm ≤0.0005
Gradient Elution Test (a.u.) - 210 nm ≤0.002
Density (g/mL) at 25°C 0.775 - 0.780
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Assay (CH₃CN) (by GC) ≥99.9 %
Appearance Passes Test
Color (APHA) ≤10
Fluorescence Trace Impurities, measured as Quinine Base - Fluorescence detection (PAH) ≤0.5 ppb
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.8
Titrable Base (µeq/g) ≤0.6
Water (by KF, coulometric) ≤100 ppm
Carbonyl Compounds (as Acetone) ≤25.0 ppb
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty