Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Methyl sulfoxideDMSODimethyl(oxido)sulfurMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphurMethanesulfinylmethane

Management of Change (MOC) category = R

BAKR9033-07 BAKR9033-QM 9033-33 BAKR9033-QM BAKR9033-04 BAKR9033-07 BAKR9033-33 BAKR9033-04
JT9033-33CS 18608.99 USD InStock
JT9033-33 CAJT9033-33 JT9033-QM CAJT9033-QM CAJT9033-07 JT9033-4 CAJT9033-4 JT9033-07
Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.45 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 89 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets U.S.P Requirements
Meets E.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Specific Gravity at 25°/25°C 1.095 - 1.101
USP - Refractive Index, n²⁵^D 1.4755 - 1.4775
USP - Acidity Passes Test
USP - Water (H₂O) ≤ 0.1 %
USP - Absorbance - 275 nm ≤ 0.20
USP - Absorbance - 270-350nm (smooth with no maxima) Passes Test
USP - Absorbance Ratio - 285 nm/275 nm ≤ 0.65
USP - Absorbance Ratio - 295 nm/275 nm ≤ 0.45
USP - Nonvolatile Residue ≤ 0.01 %
USP - Related Compounds ≤ 0.1 %
USP - Assay ((CH₃)₂SO) ≥ 99.9 %
EP - Identification Passes Test
EP - Acidity Passes Test
EP - Relative Density 1.100 - 1.104
EP - Refractive Index 1.478 - 1.480
EP - Freezing Point ≥ 18.3 °C
EP - Absorbance - 275 nm ≤ 0.30
EP - Absorbance - 285 nm ≤ 0.20
EP - Absorbance - 295 nm ≤ 0.20
EP - Absorbance - 270-350nm (smooth with no maxima) Passes Test
EP - Related Substances: Unspecified impurities, Each ≤ 0.10 %
EP - Related Substances-Total Impurities ≤ 0.15 %
EP - Water (H₂O) ≤ 0.2 %
Water (H₂O), For Information Only
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Metallic Residues: No metal catalysts or metal reagents, as defined by
EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of
this material.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty