Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

Management of Change (MOC) category = N

BAKR9070-97 9070-07 BAKR9070-01 BAKR9070-W2 BAKR9070-3C BAKR9070-97 BAKR9070-R BAKR9070-62 BAKR9070-J4 BAKR9070-W2 BAKR9070-62 BAKR9070-33 BAKR9070-3C BAKR9070-05 BAKR9070-01 BAKR9070-33 BAKR9070-98 BAKR9070-R BAKR9070-03 BAKR9070-05 BAKR9070-68 BAKR9070-68 BAKR9070-J4 BAKR9070-07 BAKR9070-98 BAKR9070-03
JT9070-7EA 474.16 USD InStock
JT9070-7 JT9070-01 CAJT9070-98 CAJT9070-1 CAJT9070-33 JT9070-68 CAJT9070-3 JT9070-3 JT9070-5 CAJT9070-97 CAJT9070-J4 JT9070-3C JT9070-W2 JT9070-R JT9070-62 CAJT9070-R CAJT9070-68 CAJT9070-W2 JT9070-97 CAJT9070-62 JT9070-98 JT9070-33 JT9070-J4 CAJT9070-3C CAJT9070-5 CAJT9070-7
Methanol ≥99.8% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC) ≥ 99.8 %
Substances Darkened by H₂SO₄ Passes Test
Substances Reducing KMnO₄ Passes Test
Solubility in H₂O Passes Test
Color (APHA) ≤ 10
Water (by KF, coulometric) ≤ 0.1 %
Residue after Evaporation ≤ 0.001 %
Carbonyl Compounds - Acetaldehyde ≤ 0.001 %
Carbonyl Compounds - Acetone ≤ 0.001 %
Carbonyl Compounds - Formaldehyde ≤ 0.001 %
Titrable Acid (µeq/g) ≤ 0.3
Titrable Base (µeq/g) ≤ 0.2
Heavy Metals (as Pb) ≤ 0.5 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty