Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

Management of Change (MOC) category = N

BAKR9084-98 BAKR9084-06 BAKR9084-03 BAKR9084-98 BAKR9084-50 BAKR9084-03 BAKR9084-01 BAKR9084-33 BAKR9084-ZR BAKR9084-05 BAKR9084-22 BAKR9084-01 BAKR9084-99 BAKR9084-99 BAKR9084-R BAKR9084-R BAKR9084-03 BAKR9084-05 BAKR9084-06 BAKR9084-07 BAKR9084-22 BAKR9084-ZR BAKR9084-50 BAKR9084-07 BAKR9084-33 BAKR9084-03
JT9084-REA 2634.51 USD InStock
JT9084-R JT9084-98 SCJT9084-3 CAJT9084-ZR JT9084-50 JT9084-99 JT9084-33 CAJT9084-22 CAJT9084-6 CAJT9084-7 CAJT9084-5 CAJT9084-3 JT9084-5 CAJT9084-1 JT9084-7 JT9084-6 TXJT90843ARR JT9084-3 CAJT9084-50 JT9084-ZR JT9084-22 CAJT9084-98 JT9084-01 CAJT9084-33 CAJT9084-99 CAJT9084-R
2-Propanol ≥99.5%, BAKER ANALYZED® ACS, J.T.Baker®
2-Propanol


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –88.5 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃CHOHCH₃) ≥ 99.5 %
Carbonyl Compounds - Acetone ≤ 0.002 %
Carbonyl Compounds - Propionaldehyde ≤ 0.002 %
Color (APHA) ≤ 10
Residue after Evaporation ≤ 0.001 %
Solubility in H₂O Passes Test
Titrable Acid or Base (meq/g) ≤ 0.0001
Water (by KF, coulometric) ≤ 0.2 %
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
ACS - Heavy Metals (as Pb) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty