Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol ≥99.9% (by GC, corrected for water content), BAKER ANALYZED™ HPLC for liquid chromatography, for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: CarbinolMethyl alcoholMethyl hydroxideMethylolWood alcohol

Filtered through a 0.2 µm filter.

J.T. Baker®
BAKR9093-5J BAKR9093-99 BAKR9093-P6 BAKR9093-60 9093-88 BAKR9093-1Z BAKR9093-2M BAKR9093-5J BAKR9093-3A BAKR9093-XN BAKR9093-6K BAKR9093-5P BAKR9093-YQ BAKR9093-3A BAKR9093-YV BAKR9093-60 BAKR9093-33 BAKR9093-XL BAKR9093-9P BAKR9093-03 BAKR9093-YV BAKR9093-98 BAKR9093-5P BAKR9093-S2 BAKR9093-P8 BAKR9093-3P BAKR9093-97 BAKR9093-9N BAKR9093-ZS BAKR9093-9N BAKR9093-8L BAKR9093-3F BAKR9093-6K BAKR9093-61 BAKR9093-48 BAKR9093-J9 BAKR9093-H2 BAKR9093-XT BAKR9093-9S BAKR9093-U5 BAKR9093-68 BAKR9093-H2 BAKR9093-XT BAKR9093-9W BAKR9093-33 BAKR9093-9S BAKR9093-50 BAKR9093-97 BAKR9093-U5 BAKR9093-3C BAKR9093-88 BAKR9093-02 BAKR9093-YZ BAKR9093-9P BAKR9093-9C BAKR9093-3H BAKR9093-ZK BAKR9093-99 BAKR9093-J9 BAKR9093-YZ BAKR9093-68 BAKR9093-76 BAKR9093-3P BAKR9093-ZK BAKR9093-S2 BAKR9093-9C BAKR9093-YQ BAKR9093-2M BAKR9093-1Z BAKR9093-YE BAKR9093-98 BAKR9093-03 BAKR9093-S1 BAKR9093-XN BAKR9093-9W BAKR9093-XL BAKR9093-3F BAKR9093-P6 BAKR9093-9V BAKR9093-76 BAKR9093-ZS BAKR9093-WK BAKR9093-2U BAKR9093-63 BAKR9093-WK BAKR9093-48 BAKR9093-3C BAKR9093-S1 BAKR9093-50 BAKR9093-61 BAKR9093-2U BAKR9093-63 BAKR9093-P8 BAKR9093-8L BAKR9093-3H BAKR9093-YE BAKR9093-9V BAKR9093-68 BAKR9093-92 BAKR9093-92
JT9093-3CS 1071.47 USD InStock
CAJT9093-76 JT9093-3 JT9093-2 JT9093-68 CAJT9093-P6 CAJT9093-H2 CAJT9093-P8 JT9093-5J CAJT9093-WK CAJT9093-6K JT9093-5P JT9093-61 JT9093-60 JT9093-63 JT9093-76 CAJT9093-XT CAJT9093-88 JT9093-P6 JT9093-P8 JT9093-H2 JT9093-6K JT9093-WK CAJT9093-XN CAJT9093-XL CAJT9093-97 CAJT9093-98 CAJT9093-YV JT9093-88 CAJT9093-99 CAJT9093-YZ CAJT9093-9C JT9093-YE CAJT9093-YE JT9093-XL JT9093-XN CAJT9093-8L CAJT9093-YQ JT9093-XT CAJT9093-92 JT9093-98 CAJT9093-9W CAJT9093-S1 JT9093-97 CAJT9093-S2 JT9093-YZ JT9093-99 CAJT9093-9V JT9093-9C CAJT9093-1Z CAJT9093-J9 CAJT9093-ZK JT9093-8L CAJT9093-9P JT9093-92 CAJT9093-9N JT9093-YQ CAJT9093-9S JT9093-YV CAJT9093-ZS JT9093-S2 JT9093-S1 JT9093-9W CAJT9093-2U CAJT9093-33 JT9093-1Z JT9093-J9 JT9093-ZK JT9093-9N JT9093-9P JT9093-ZS JT9093-9S CAJT9093-2M JT9093-9V JT9093-33 CAJT9093-3P JT9093-2U CA35001-236 CAJT9093-U5 CAJT9093-48 CAJT9093-3A CAJT9093-3F CAJT9093-3C CAJT9093-3H JT9093-2M CAJT9093-3 JT9093-U5 JT9093-48 JT9093-3A JT9093-3C JT9093-3F JT9093-3H CAJT9093-50 JT9093-3P CAJT9093-63 CAJT9093-68 CAJT9093-5J CAJT9093-60 JT9093-50 CAJT9093-5P CAJT9093-61
Methanol ≥99.9% (by GC, corrected for water content), BAKER ANALYZED™ HPLC for liquid chromatography, for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Methanol


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (CH₃OH) (by GC, corrected for water) ≥99.9 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.15
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Gradient Elution Test (a.u.) - 254 nm ≤0.002
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.3 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤1.0 ppb
Acetone ≤0.001 %
Residue after Evaporation ≤1.0 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.10
Water (by KF, coulometric) ≤0.05 %
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty