Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Propanol ≥99.7%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Isopropyl alcoholIsopropyl Rubbing Alcoholsec-PropanolIsopropyl rubbing alcohol2-Hydroxy propanesec-Propyl AlcoholDimethyl carbinol2-HydroxypropanePropane, 2-hydroxy-Propan-2-olIPAIsopropanol

Management of Change (MOC) category = N

BAKR9095-02 BAKR9095-03 9095-88 BAKR9095-99 BAKR9095-3H BAKR9095-03 BAKR9095-6K BAKR9095-33 BAKR9095-6K BAKR9095-33 BAKR9095-88 BAKR9095-02 BAKR9095-03 BAKR9095-3H BAKR9095-99
JT9095-99CS 3383 USD InStock
CAJT9095-88 CAJT9095-33 CAJT9095-99 JT9095-99 JT9095-33 JT9095-88 JT9095-3 JT9095-2 TXLJT9095-3GSK CAJT9095-3H CAJT9095-6K JT9095-6K JT9095-3H CAJT9095-2 CAJT9095-3
2-Propanol ≥99.7%, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
2-Propanol


Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.786 g/cm³ (20 °C)
Flash Pt: 12 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3,II
REACH: 01-2119457558-25
Merck Index: 13,05228
https://us-legacy.vwr.com/stibo/web/std.lang.all/19/69/731969.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm ≤0.020
Ultraviolet Absorbance (1.00-cm cell vs. water) - 225 nm ≤0.16
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤205
Assay (CH₃CHOHCH₃) ≥99.7 %
Residue after Evaporation ≤2.0 ppm
Water (by KF, coulometric) ≤0.03 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty