Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorobenzene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Phenylchloride

Management of Change (MOC) category = N

BAKR9179-08 BAKR9179-50 BAKR9179-01 BAKR9179-50 BAKR9179-03 BAKR9179-99 BAKR9179-98 BAKR9179-99 BAKR9179-01 BAKR9179-03 BAKR9179-98 BAKR9179-08
JT9179-99CS 4475.19 USD InStock
CAJT9179-3 JT9179-99 JT9179-98 CAJT9179-99 CAJT9179-8 CAJT9179-98 JT9179-50 JT9179-3 JT9179-1 CAJT9179-50 JT9179-8 CAJT9179-1
Chlorobenzene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Chlorobenzene


Formula: C₆H₅Cl
MW: 112.56 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: –45 °C
Density: 1.11 g/cm³ (20 °C)
Flash Pt: 28 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00000530
CAS Number: 108-90-7
EINECS: 203-628-5
UN: 1134
ADR: 3,III
REACH: 01-2119432722-45
Merck Index: 13,02139
https://us-legacy.vwr.com/stibo/web/std.lang.all/08/56/750856.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₆H₅Cl) (by GC) ≥99.5 %
ACS - Acidity (as HCl) ≤0.015 %
ACS - Color (APHA) ≤30
ACS - Residue after Evaporation ≤0.010 %
ACS - Titrable Acid (meq/g) ≤0.004

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty