Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with ethanol 1 %

BAKR9180-50 BAKR9180-98 BAKR9180-05 BAKR9180-01 BAKR9180-R BAKR9180-08 BAKR9180-22 BAKR9180-05 BAKR9180-99 BAKR9180-98 BAKR9180-33 BAKR9180-R BAKR9180-54 BAKR9180-J6 BAKR9180-22 BAKR9180-03 BAKR9180-33 BAKR9180-08 BAKR9180-01 BAKR9180-03 BAKR9180-99 BAKR9180-50 BAKR9180-J6
JT9180-01CS 1543.94 USD InStock
CAJT9180-99 CAJT9180-33 CAJT9180-22 CAJT9180-98 CAJT9180-54 JT9180-01 JT9180-22 JT9180-33 JT9180-99 JT9180-98 CAJT9180-50 JT9180-J6 CAJT9180-5 JT9180-R CAJT9180-8 CAJT9180-1 JT9180-5 CAJT9180-R CAJT9180-3 JT9180-3 JT9180-8 CAJT9180-J6 JT9180-50
Chloroform ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 61.7 °C (1013 hPa)
Melting Pt: –63 °C
Density: 1.4832 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water) ≥99.8 %
Acetone, Aldehydes (as (CH₃)₂CO) Passes Test
Acid and Chloride Passes Test
Color (APHA) ≤10
Ethanol (CH₃CH₂OH)(by GC) 0.7 - 1.0 %
Free Chlorine Passes Test
Lead (Pb) ≤0.050 ppm
Residue after Evaporation ≤2.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Suitability for Use in Dithizone Tests Passes Test
Water (by KF, coulometric) ≤0.01 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty