Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Methyl sulfoxideDMSOMethyl sulphoxide

9194-03 BAKR9194-03 BAKR9194-33 9194-33
35001-238CS 2540.57 USD InStock
35001-238 CAJT9194-33 JT9194-33 CAJT9194-3
Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Dimethyl sulphoxide
For spectrophotometry. UV cutoff 270 nm. Lot analysis on label.


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C (closed cup)
MDL Number: MFCD00002089
CAS Number: 67-68-5
ADR: 4,
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
ACS - Assay ((CH₃)₂SO) (by GC, corrected for water) ≥99.9 %
ACS - Residue after Evaporation ≤0.002 %
ACS - Titrable Acid (meq/g) ≤0.001
ACS - Water (by KF, coulometric) ≤0.1 %
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400-350 ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 330 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 310 nm ≤0.06
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 290 nm ≤0.18
Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm ≤0.40
Product Information (not specifications):
Windows of Infrared Transmittance (0.1-mm path, 50-100% T), µm:
2.0 - 2.3; 2.4 - 3.2; 3.7 – 5.0; 5.1 – 6.7; 7.9 – 8.7; 11.4 – 13.8

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty