Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKR9210-01 BAKR9210-01 BAKR9210-03 BAKR9210-03
JT9210-3CS 1443.46 USD InStock
CAJT9210-3 CAJT9210-1 JT9210-3 JT9210-1
Cyclohexanone ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Cyclohexanone


Formula: C₆H₁₀(=O)
MW: 98.14 g/mol
Boiling Pt: 155.6 °C (1013 hPa)
Melting Pt: –31 °C
Density: 0.9478 g/cm³ (20 °C)
Flash Pt: 43 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001625
CAS Number: 108-94-1
EINECS: 203-631-1
UN: 1915
ADR: 3,III
REACH: 01-2119453616-35
Merck Index: 13,02755
https://us-legacy.vwr.com/stibo/web/std.lang.all/12/57/771257.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₁₀O) (by GC) ≥99.0 %
Color (APHA) ≤10
Density (g/mL) at 25°C 0.940 - 0.943
Residue after Evaporation ≤0.0500 %
Acidity (meq/g) ≤0.003
Water (H₂O)(by Karl Fischer titrn) ≤0.1 %
Identification (by IR) Passes Test
NOTE: Color reported is at time of packaging; material will darken with age.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty