Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: NSC 5356Formic acid dimethylamideDimethylformamideDMFADMFFormyldimethylamineN-Formyldimethylamine

Management of Change (MOC) category = N

BAKR9221-01 BAKR9221-33 BAKR9221-98 BAKR9221-06 9221-R BAKR9221-33 BAKR9221-98 BAKR9221-01 BAKR9221-03 BAKR9221-06 BAKR9221-03 BAKR9221-99 BAKR9221-99 BAKR9221-R
JT9221-6CS 1828.16 USD InStock
CAJT9221-3 CAJT9221-R CAJT9221-1 CAJT9221-98 CAJT9221-33 JT9221-6 CAJT9221-99 JT9221-3 JT9221-1 JT9221-R JT9221-33 JT9221-99 JT9221-98 CAJT9221-6
N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
N,N-Dimethylformamide


Formula: HCON(CH₃)₂
MW: 73.09 g/mol
Boiling Pt: 153 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.9445 g/cm³ (25 °C)
Flash Pt: 57.5 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003284
CAS Number: 68-12-2
EINECS: 200-679-5
UN: 2265
ADR: 3,III
REACH: 01-2119475605-32
Merck Index: 13,03269
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/06/2302506.jpg https://us-legacy.vwr.com/stibo/web/std.lang.all/06/12/16640612.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (HCON(CH₃)₂) (by GC, corrected for water) ≥ 99.8 %
Appearance Passes Test
Color (APHA) ≤ 15
Residue after Evaporation ≤ 0.0050 %
Titrable Acid (meq/g) ≤ 0.0005
Titrable Base (meq/g) ≤ 0.003
Water (H₂O)(by Karl Fischer titrn) ≤ 0.1 %
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty