Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Methyl sulfoxideDMSODimethyl(oxido)sulfurMethyl sulphoxideMethanesulphinylmethaneDimethyl(oxido)sulphurMethanesulfinylmethane

Management of Change (MOC) category = N

BAKR9224-07 BAKR9224-99 BAKR9224-06 BAKR9224-70 BAKR9224-06 BAKR9224-76 BAKR9224-01 BAKR9224-50 BAKR9224-01 BAKR9224-33 BAKR9224-76 BAKR9224-50 BAKR9224-72 BAKR9224-33 BAKR9224-98 BAKR9224-03 BAKR9224-03 BAKR9224-72 BAKR9224-70 BAKR9224-98 BAKR9224-07 BAKR9224-61 BAKR9224-99 BAKR9224-61
JT9224-6CS 4471.71 USD InStock
JT9224-6 CAJT9224-76 CAJT9224-1 CAJT9224-98 JT9224-33 JT9224-7 JT9224-99 CAJT9224-99 CAJT9224-33 JT9224-3 JT9224-1 JT9224-70 CAJT9224-7 JT9224-50 JT9224-72 CAJT9224-70 JT9224-61 CAJT9224-6 CAJT9224-50 CAJT9224-72 CAJT9224-61 JT9224-76 CAJT9224-3 JT9224-98
Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay ((CH₃)₂SO) (by GC, corrected for water) ≥99.9 %
Residue after Evaporation ≤0.0100 %
Titrable Acid (meq/g) ≤0.001
Water (by KF, coulometric) ≤0.1 %
Product Information (not specifications):
Density (g/mL) at 25°C (typical) = 1.092
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty