Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

BAKR9280-05 BAKR9280-ZX 9280-02 BAKR9280-33 BAKR9280-98 BAKR9280-7R BAKR9280-99 BAKR9280-03 BAKR9280-99 BAKR9280-J6 BAKR9280-03 BAKR9280-J6 BAKR9280-07 BAKR9280-01 BAKR9280-22 BAKR9280-ZX BAKR9280-01 BAKR9280-98 BAKR9280-22 BAKR9280-07 BAKR9280-7R BAKR9280-05 BAKR9280-3C BAKR9280-R BAKR9280-3C BAKR9280-33
JT9280-3CS 745.21 USD InStock
JT9280-3 40004-726 JT9280-5 JT9280-7 CAJT9280-3 JT9280-7R CAJT9280-1 JT9280-3C CAJT9280-33 CAJT9280-99 JT9280-01 JT9280-22 JT9280-ZX CAJT9280-98 CAJT9280-7R CAJT9280-R CAJT9280-3C CAJT9280-J6 JT9280-J6 CAJT9280-22 JT9280-99 CAJT9280-7 JT9280-33 JT9280-98 CAJT9280-5 CAJT9280-ZX
Ethyl acetate ≥99.9% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Ethyl acetate


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₃COOC₂H₅) (by GC, corrected for water) ≥99.9 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Titrable Acid (µeq/g) ≤0.9
Water (by KF, volumetric) ≤0.06 %
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty