Synonyms:
Reagent alcohol
Management of Change (MOC) category = N
BAKR9287-03
BAKR9287-07
BAKR9287-07
BAKR9287-33
BAKR9287-33
BAKR9287-03
JT9287-33CS
1145.98
USD
InStock
CAJT9287-7
CAJT9287-33
JT9287-33
JT9287-3
CAJT9287-3
JT9287-7
Alcohol, Proprietary Solvent III-1, anhydrous, BAKER ANALYZED®, J.T.Baker®
Alcohol
Denatured with 1% ethyl acetate, 1% methyl isobutyl ketone, 1% gasoline, and 4 to 6% methanol. Lot analysis on label.
- Total alcohol: 99.5%
- Ethyl alcohol: 96%
- Denaturant: methanol and a small amount of denatonium benzoate
SDA Formula 1-1 consists of 4 volumes of methanol, 1/8 oz of denatonium benzoate, and 100 volumes of 200 proof ethanol. * Specially Denatured Alcohol Formula 1-1 (200 Proof)(v/v)(parts by vol.)........ 100 *
Ethyl Acetate (v/v)(parts by vol)....... 0.87 * Methyl Isobutyl Ketone (v/v)(parts by vol)..... 1 * Rubber Hydrocarbon Solvent (v/v)(parts by vol)... 1