Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: 1,2-Ethanediol1,2-Dihydroxyethane1,2-Dihydroxy ethaneEthanediolGlycol alcoholDihydroxyethaneEthylene alcoholEthane-1,2-diolMonoethylene glycol2-HydroxyethanolGlycol

BAKR9300-R BAKR9300-07 BAKR9300-01 BAKR9300-03 BAKR9300-01 BAKR9300-R BAKR9300-01 BAKR9300-07 BAKR9300-33 BAKR9300-03 BAKR9300-33 BAKR9300-03
JT9300-3CS 2146.21 USD InStock
JT9300-3 JT9300-R CAJT9300-1 CAJT9300-R JT9300-7 CAJT9300-7 JT9300-33 JT9300-01 CAJT9300-33 SCJT9300-1 CAJT9300-3 SCJT9300-3
Ethylene glycol ≥99.0% (by GC), BAKER ANALYZED®, J.T.Baker®
Ethylene glycol


Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/75/734375.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (HOCH₂CH₂OH) (by GC) ≥99 %
Assay (HOCH₂CH₂OH)(wet analysis) ≥99 %
Color (APHA) ≤10
Specific Gravity at 20°/20°C 1.114 - 1.116
Residue after Ignition ≤0.005 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty