Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with cyclohexene ≥ 50 ppm
Management of Change (MOC) category = N

BAKR9315-Q9 BAKR9315-Q8 BAKR9315-99 BAKR9315-Q6 BAKR9315-33 BAKR9315-Q7 BAKR9315-S8 BAKR9315-98 BAKR9315-Q8 BAKR9315-4C BAKR9315-Q9 BAKR9315-6Q BAKR9315-S8 BAKR9315-03 BAKR9315-02 BAKR9315-4C BAKR9315-6Q BAKR9315-Q6 BAKR9315-99 BAKR9315-50 BAKR9315-98 BAKR9315-50 BAKR9315-03 BAKR9315-Q7
JT9315-6QCS 4030.32 USD InStock
JT9315-6Q JT9315-50 JT9315-2 CAJT9315-6Q JT9315-3 CAJT9315-50 JT9315-98 JT9315-33 JT9315-99 CAJT9315-98 CAJT9315-99 CAJT9315-3 JT9315-Q6 JT9315-S8 JT9315-Q7 JT9315-Q8 CAJT9315-S8 CAJT9315-Q6 JT9315-Q9 CAJT9315-Q7 CAJT9315-Q8 CAJT9315-Q9 JT9315-4C CAJT9315-4C
Dichloromethane ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Dichloromethane
Methylene Chloride HPLC For use in liquid chromatography (HPLC and UHPLC) and spectrophotometry.


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-254 nm ≤0.01
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission For Information Only
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission For Information Only
Fluorescence Trace Impurities measured as Quinine Base at Emission Maximum for Impurities, For Information Only
Maximum for Impurities, For Information Only
Assay (by GC, corrected for water, exclusive of preservative) ≥99.8 %
Chloride (Cl) ≤10 ppm
Preservative (cyclohexene) ≥50 ppm
Residue after Evaporation ≤2.0 ppm
Titrable Acid (µeq/g) ≤0.3
Water (by KF, coulometric) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty