Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methane dichlorideMethylene chloride

BAKR9324-98 BAKR9324-07 BAKR9324-98 BAKR9324-ZK BAKR9324-22 BAKR9324-99 BAKR9324-ZR BAKR9324-R BAKR9324-01 BAKR9324-99 BAKR9324-W2 BAKR9324-W2 BAKR9324-07 9324-03 BAKR9324-05 BAKR9324-ZR BAKR9324-ZX BAKR9324-33 BAKR9324-ZK BAKR9324-50 BAKR9324-R BAKR9324-ZX BAKR9324-01 BAKR9324-03 BAKR9324-03 BAKR9324-05 BAKR9324-22 BAKR9324-50 BAKR9324-33
JT9324-3CS 2518.56 USD InStock
JT9324-3 JT9324-5 CAJT9324-98 JT9324-01 CAJT9324-99 CAJT9324-33 JT9324-22 CAJT9324-5 CAJT9324-50 CAJT9324-7 JT9324-ZX JT9324-W2 CAJT9324-1 CAJT9324-3 CAJT9324-ZK SCJT9324-3 JT9324-7 JT9324-R CAJT9324-22 JT9324-98 JT9324-99 CAJT9324-ZX JT9324-33 CAJT9324-W2 CAJT9324-R JT9324-50 CAJT9324-ZR JT9324-ZR JT9324-ZK
Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (CH₂Cl₂) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.0020 %
Titrable Acid (µeq/g) ≤0.3
Free Halogens Passes Test
Water (by KF, coulometric) ≤0.02 %
Product Information (not specifications):
Appearance (Clear, colorless liquid)
Boiling Point (typical)................. 39.5 °C.
Density (g/mL) at 25 °C (typical)........ 1.318
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty