Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: MethacideBenzene, methylMethylbenzolToluolPhenylmethaneMethylbenzeneMethane, phenyl-

Management of Change (MOC) category = N

BAKR9351-33 BAKR9351-03 BAKR9351-33 BAKR9351-03 BAKR9351-02 BAKR9351-02
JT9351-3CS 977.25 USD InStock
CAJT9351-3 CAJT9351-2 JT9351-3 CAJT9351-33 JT9351-33 JT9351-2
Toluene ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Toluene
  • UV cutoff 285 nm


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (by GC, corrected for water) ≥99.7 %
Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm ≤0.10
Ultraviolet Absorbance (1.00-cm cell vs. water) - 288 nm ≤0.40
Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm ≤285
Residue after Evaporation ≤3.0 ppm
Substances Darkened by H₂SO₄ Passes Test
Water (by KF, coulometric) ≤0.02 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty