Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Benzene, methylMethacideMethane, phenyl-MethylbenzeneMethylbenzolPhenylmethaneToluol

BAKR9460-61 BAKR9460-9K BAKR9460-Q8 BAKR9460-Q6 BAKR9460-03 BAKR9460-07 BAKR9460-8Z BAKR9460-33 BAKR9460-8Z BAKR9460-05 BAKR9460-Q8 BAKR9460-50 BAKR9460-07 BAKR9460-9K BAKR9460-4C BAKR9460-01 BAKR9460-03 BAKR9460-Q6 BAKR9460-98 BAKR9460-4C BAKR9460-05 BAKR9460-33 BAKR9460-98 BAKR9460-50 BAKR9460-Q7 BAKR9460-61 BAKR9460-22 BAKR9460-S8 BAKR9460-S8 BAKR9460-Q7 BAKR9460-R BAKR9460-R BAKR9460-22
JT9460-4CCS 5685.86 USD InStock
JT9460-4C CAJT9460-33 JT9460-Q6 JT9460-S8 CAJT9460-8Z JT9460-Q8 JT9460-Q7 CAJT9460-5 JT9460-61 CAJT9460-50 CAJT9460-7 JT9460-7 CAJT9460-3 JT9460-01 CAJT9460-98 JT9460-22 JT9460-9K JT9460-3 JT9460-5 CAJT9460-4C JT9460-8Z CAJT9460-22 CAJT9460-Q6 CAJT9460-S8 CAJT9460-Q8 CAJT9460-Q7 JT9460-50 CAJT9460-61 JT9460-98 JT9460-33 CAJT9460-R JT9460-R CAJT9460-9K
Toluene ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Toluene


Formula: C₆H₅CH₃
MW: 92.14 g/mol
Boiling Pt: 110.6 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00008512
CAS Number: 108-88-3
EINECS: 203-625-9
UN: 1294
ADR: 3,II
REACH: 05-2114615130-69
Merck Index: 12,09667
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/73/766173.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (C₆H₅CH₃) (by GC) ≥99.5 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Substances Darkened by H₂SO₄ Passes Test
Sulfur Compounds (as S) ≤0.003 %
Water (by KF, coulometric) ≤0.03 %
Date Received: __ __ /__ __ /__ __
Date Opened: __ __ /__ __ /__ __
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty