Synonyms:
Methanecarboxylic acid
, Vinegar acid
, Ethanoic acid
Management of Change (MOC) category = N
BAKR9508-03
BAKR9508-00
BAKR9508-00
BAKR9508-07
BAKR9508-15
BAKR9508-01
BAKR9508-69
BAKR9508-33
BAKR9508-08
BAKR9508-33
BAKR9508-05
BAKR9508-02
BAKR9508-15
BAKR9508-18
BAKR9508-08
BAKR9508-06
BAKR9508-06
BAKR9508-03
BAKR9508-00
9508-07
BAKR9508-18
9508-69
BAKR9508-01
SCJT9508-0CS
443.01
USD
InStock
SCJT9508-0
JT9508-33
SCJT9508-3
CAJT9508-0
CAJT9508-1
JT9508-7
JT9508-8
CAJT9508-18
CAJT9508-6
CAJT9508-7
CAJT9508-8
CAJT9508-15
JT9508-0
CAJT9508-69
JT9508-1
JT9508-2
JT9508-18
JT9508-3
CAJT9508-33
JT9508-5
JT9508-15
JT9508-69
JT9508-6
Acetic acid glacial ≥99.7% (by GC, corrected for water content), BAKER ANALYZED® ACS aldehyde-free, J.T.Baker®
Acetic acid
The quality of BAKER ANALYZED™ ACS Reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.
Caution: IMPORTANT: Material will freeze if stored below 17 °C (63 °F).