Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

Management of Change (MOC) category = N

BAKR9511-05 BAKR9511-02 BAKR9511-05
JT9511-2CS 1775 USD InStock
JT9511-2 JT9511-5 CAJT9511-5
Acetic acid glacial ≥99.7%, BAKER ANALYZED® ACS, J.T.Baker®
Acetic acid
For cholesterol determination. Aldehyde-free. Lot analysis on label.


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 38.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
REACH: 01-2119475328-30
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
IMPORTANT: Material will freeze if stored below 17 °C (63°F).
ACS - Assay (by GC, corrected for water) ≥99.7 %
ACS - Color (APHA) ≤10
Dilution Test Passes Test
ACS - Residue after Evaporation ≤0.001 %
ACS - Acetic Anhydride ((CH₃CO)₂O) ≤0.01 %
ACS - Chloride (Cl) ≤1 ppm
ACS - Sulfate (SO₄) ≤1 ppm
ACS - Substances Reducing Dichromate Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Base (meq/g) ≤0.0004
ACS - Suitability for Cholesterol Determination Passes Test
ACS - Acetaldehyde ≤0.005 %
ACS - Heavy Metals (by ICP-OES) ≤5 ppm
Trace Impurities - ACS - Iron (Fe) ≤0.200 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty