Specification Test Results
For Laboratory,Research,or Manufacturing Use |
|
IMPORTANT: Material will freeze if stored below 17 °C (63°F). |
|
ACS - Assay (by GC, corrected for water) |
≥99.9 % |
ACS - Acetaldehyde |
≤0.005 % |
ACS - Acetic Anhydride ((CH₃CO)₂O) |
≤0.01 % |
ACS - Color (APHA) |
≤10 |
Dilution Test |
Passes Test |
ACS - Residue after Evaporation |
≤5 ppm |
ACS - Solubility in H₂O |
Passes Test |
ACS - Specific Gravity at 20°/20°C |
≥1.048 |
ACS - Substances Reducing Dichromate |
Passes Test |
ACS - Substances Reducing Permanganate |
Passes Test |
ACS - Titrable Base (meq/g) |
≤0.0004 |
Chloride (Cl) |
≤0.5 ppm |
Phosphate (PO₄) |
≤1 ppm |
Sulfate (SO₄) |
≤0.5 ppm |
Trace Impurities - Aluminum (Al) |
≤50.0 ppb |
Arsenic and Antimony (as As) |
≤5.0 ppb |
Trace Impurities - Barium (Ba) |
≤20.0 ppb |
Trace Impurities - Beryllium (Be) |
≤10.0 ppb |
Trace Impurities - Bismuth (Bi) |
≤50 ppb |
Trace Impurities - Boron (B) |
≤10.0 ppb |
Trace Impurities - Cadmium (Cd) |
≤5.0 ppb |
Trace Impurities - Calcium (Ca) |
≤100.0 ppb |
Trace Impurities - Chromium (Cr) |
≤50 ppb |
Trace Impurities - Cobalt (Co) |
≤10.0 ppb |
Trace Impurities - Copper (Cu) |
≤10.0 ppb |
Trace Impurities - Germanium (Ge) |
≤50.0 ppb |
Heavy Metals (as Pb) |
≤300.0 ppb |
Trace Impurities - Iron (Fe) |
≤50.0 ppb |
Trace Impurities - Lead (Pb) |
≤10.0 ppb |
Trace Impurities - Lithium (Li) |
≤50.0 ppb |
Trace Impurities - Magnesium (Mg) |
≤50.0 ppb |
Trace Impurities - Manganese (Mn) |
≤5.0 ppb |
Trace Impurities - Mercury (Hg) |
≤5.0 ppb |
Trace Impurities - Nickel (Ni) |
≤10.0 ppb |
Trace Impurities - Potassium (K) |
≤200.0 ppb |
Trace Impurities - Silicon (Si) |
≤50 ppb |
Trace Impurities - Silver (Ag) |
≤5.0 ppb |
Trace Impurities - Sodium (Na) |
≤500.0 ppb |
Trace Impurities - Strontium (Sr) |
≤5.0 ppb |
Trace Impurities - Tin (Sn) |
≤10.0 ppb |
Trace Impurities - Titanium (Ti) |
≤300.0 ppb |
Trace Impurities - Zinc (Zn) |
≤10.0 ppb |