Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Muriatic acid

Management of Change (MOC) category = R

BAKR9535-33 9535-69 BAKR9535-19 BAKR9535-15 BAKR9535-07 BAKR9535-33 BAKR9535-A1 BAKR9535-08 BAKR9535-A1 BAKR9535-03 BAKR9535-03 BAKR9535-33 BAKR9535-00 BAKR9535-09 BAKR9535-01 BAKR9535-00 9535-05 BAKR9535-02 BAKR9535-15 BAKR9535-08 BAKR9535-07 BAKR9535-69 BAKR9535-03 BAKR9535-19
JT9535-0CS 451.88 USD InStock
CAJT9535-33 JT9535-0 JT9535-33 JT9535-2 JT9535-1 JT9535-3 CAJT9535-19 JT9535-19 CAJT9535-0 CAJT9535-69 JT9535-05 CAJT9535-15 JT9535-69 CAJT9535-3 JT9535-15 CAJT9535-9 SCJT9535-33 CAJT9535-A1 CAJT9535-7 CAJT9535-8 JT9535-A1 JT9535-8 JT9535-7 SCJT9535-3
Hydrochloric acid 36.5 - 38.0%, BAKER ANALYZED® ACS, J.T.Baker®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Store below 25°C
Appearance Passes Test
Assay (as HCl) (by acid-base titrn) 36.5 - 38.0 %
Color (APHA) ≤ 10
Extractable Organic Substances ≤ 5 ppm
Free Chlorine (as Cl₂) ≤ 1 ppm
Residue after Ignition ≤ 3 ppm
Specific Gravity at 60°/60°F 1.185 - 1.192
Bromide (Br) ≤ 0.005 %
Phosphate (PO₄) ≤ 1 ppm
Sulfate (SO₄) ≤ 0.5 ppm
Sulfite (SO₃) ≤ 0.8 ppm
Ammonium (NH₄) ≤ 3 ppm
Trace Impurities - Aluminum (Al) ≤ 100.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Boron (B) ≤ 50 ppb
Trace Impurities - Calcium (Ca) ≤ 200.0 ppb
Trace Impurities - Chromium (Cr) ≤ 100.0 ppb
Trace Impurities - Copper (Cu) ≤ 100.0 ppb
Trace Impurities - Gold (Au) ≤ 100.0 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 100.0 ppb
Trace Impurities - Lead (Pb) ≤ 50 ppb
Trace Impurities - Magnesium (Mg) ≤ 300.0 ppb
Trace Impurities - Manganese (Mn) ≤ 300.0 ppb
Trace Impurities - Mercury (Hg) ≤ 5.0 ppb
Trace Impurities - Nickel (Ni) ≤ 100.0 ppb
Trace Impurities - Potassium (K) ≤ 300.0 ppb
Trace Impurities - Sodium (Na) ≤ 300 ppb
Trace Impurities - Tin (Sn) ≤ 300.0 ppb
Trace Impurities - Titanium (Ti) ≤ 300.0 ppb
Trace Impurities - Zinc (Zn) ≤ 100.0 ppb
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty