Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 37.0 - 38.0%, CMOS for microelectronic, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

Management of Change (MOC) category = R

9539-05 BAKR9539-01 9539-03 BAKR9539-05 BAKR9539-33 BAKR9539-03 BAKR9539-33
JT9539-33CS 2173.48 USD InStock
CAJT9539-5 CAJT9539-3 JT9539-33 JT9539-01 JT9539-3 CAJT9539-33 JT9539-5
Hydrochloric acid 37.0 - 38.0%, CMOS for microelectronic, J.T.Baker®
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
For Microelectronic Use
Store below 25°C
Assay (as HCl) (by acid-base titrn) 37.0 - 38.0 %
Color (APHA) ≤ 10
Residue after Ignition ≤ 3 ppm
Extractable Organic Substances ≤ 3 ppm
Bromide (Br) ≤ 0.005 %
Free Halogen (as Cl₂) Passes Test
Ammonium (NH₄) ≤ 1 ppm
Phosphate (PO₄) ≤ 0.05 ppm
Sulfate (SO₄) ≤ 0.3 ppm
Sulfite (SO₃) ≤ 0.8 ppm
Trace Impurities - Aluminum (Al) ≤ 50.0 ppb
Arsenic and Antimony (as As) ≤ 5.0 ppb
Trace Impurities - Barium (Ba) ≤ 20.0 ppb
Trace Impurities - Beryllium (Be) ≤ 10.0 ppb
Trace Impurities - Bismuth (Bi) ≤ 20.0 ppb
Trace Impurities - Boron (B) ≤ 20.0 ppb
Trace Impurities - Cadmium (Cd) ≤ 5.0 ppb
Trace Impurities - Calcium (Ca) ≤ 100.0 ppb
Trace Impurities - Chromium (Cr) ≤ 10.0 ppb
Trace Impurities - Cobalt (Co) ≤ 5.0 ppb
Trace Impurities - Copper (Cu) ≤ 5.0 ppb
Trace Impurities - Gallium (Ga) ≤ 20.0 ppb
Trace Impurities - Germanium (Ge) ≤ 20 ppb
Trace Impurities - Gold (Au) ≤ 20 ppb
Heavy Metals (as Pb) ≤ 100 ppb
Trace Impurities - Iron (Fe) ≤ 50.0 ppb
Trace Impurities - Lead (Pb) ≤ 20.0 ppb
Trace Impurities - Lithium (Li) ≤ 20.0 ppb
Trace Impurities - Magnesium (Mg) ≤ 50.0 ppb
Trace Impurities - Manganese (Mn) ≤ 5.0 ppb
Trace Impurities - Molybdenum (Mo) ≤ 10.0 ppb
Trace Impurities - Nickel (Ni) ≤ 10.0 ppb
Trace Impurities - Niobium (Nb) ≤ 10.0 ppb
Trace Impurities - Potassium (K) ≤ 50 ppb
Trace Impurities - Silicon (Si) ≤ 100.0 ppb
Trace Impurities - Silver (Ag) ≤ 20.0 ppb
Trace Impurities - Sodium (Na) ≤ 100.0 ppb
Trace Impurities - Strontium (Sr) ≤ 20.0 ppb
Trace Impurities - Tantalum (Ta) ≤ 10.0 ppb
Trace Impurities - Thallium (Tl) ≤ 20.0 ppb
Trace Impurities - Tin (Sn) ≤ 50 ppb
Trace Impurities - Titanium (Ti) ≤ 10.0 ppb
Trace Impurities - Vanadium (V) ≤ 10.0 ppb
Trace Impurities - Zinc (Zn) ≤ 30.0 ppb
Trace Impurities - Zirconium (Zr) ≤ 10.0 ppb
Particle Count - 0.5 µm and greater ≤ 50 par/ml
Particle Count - 1.0 µm and greater ≤ 10 par/ml

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty