Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitric acid 69.0 - 70.0% NF, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Nitric acid solution

Management of Change (MOC) category = R

9607-33 BAKR9607-3X BAKR9607-04 BAKR9607-3X BAKR9607-33
JT9607-4CS 802.16 USD InStock
JT9607-4 CAJT9607-33 CAJT9607-3X JT9607-3X JT9607-33
Nitric acid 69.0 - 70.0% NF, J.T.Baker®
Nitric acid
Corrosive.


Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 120.5 °C (1013 hPa)
Melting Pt: –42 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (as HNO₃) 69.0 - 70.0 %
NF - Clarity and Color Passes Test
NF - Identification Passes Test
NF - Chloride (Cl) ≤0.5 ppm
NF - Sulfate (SO₄) ≤1 ppm
NF - Iron (Fe) ≤0.2 ppm
NF - Residue on Ignition ≤5 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty