Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95.0-98.0% (by acidimetry) NF, ACS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

J.T. Baker®
BAKR9661-03 BAKR9661-03
JT9661-3CS 584.51 USD InStock
JT9661-3 CAJT9661-3
Sulfuric acid 95.0-98.0% (by acidimetry) NF, ACS, J.T.Baker®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
GMP Manufactured Product
Meets N.F. Requirements
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Assay (H₂SO₄) (by acidimetry) 95.0 - 98.0 %
NF - Arsenic (As) ≤1 ppm
NF - Chloride (Cl) ≤0.005 %
NF - Identification Passes Test
NF - Reducing Substances Passes Test
NF - Residue on Ignition ≤0.005 %
Ammonium (NH₄) ≤1 ppm
Appearance Passes Test
Assay (H₂SO₄) 95.0 - 98.0 %
Chloride (Cl) ≤0.1 ppm
Color (APHA) ≤10
Heavy Metals (as Pb) ≤500.0 ppb
Nitrate (NO₃) ≤0.5 ppm
Residue after Ignition ≤4 ppm
Specific Gravity at 60°/60°F ≥1.84
Substances Reducing Permanganate (as SO₂) ≤2 ppm
Trace Impurities - Aluminum (Al) ≤200.0 ppb
Trace Impurities - Arsenic (As) ≤0.004 ppm
Trace Impurities - Calcium (Ca) ≤200.0 ppb
Trace Impurities - Chromium (Cr) ≤100.0 ppb
Trace Impurities - Copper (Cu) ≤100.0 ppb
Trace Impurities - Gold (Au) ≤100.0 ppb
Trace Impurities - Iron (Fe) ≤200.0 ppb
Trace Impurities - Lead (Pb) ≤200.0 ppb
Trace Impurities - Magnesium (Mg) ≤200.0 ppb
Trace Impurities - Manganese (Mn) ≤100.0 ppb
Trace Impurities - Mercury (Hg) ≤5.0 ppb
Trace Impurities - Nickel (Ni) ≤200.0 ppb
Trace Impurities - Potassium (K) ≤300.0 ppb
Trace Impurities - Sodium (Na) ≤300 ppb
Trace Impurities - Tin (Sn) ≤200.0 ppb
Trace Impurities - Titanium (Ti) ≤200.0 ppb
Trace Impurities - Zinc (Zn) ≤200.0 ppb
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty