Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetonitrile with 0.1% (v/v) trifluoroacetic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger

Management of Change (MOC) category = NI

BAKR9835-02 BAKR9835-03 BAKR9835-02 BAKR9835-03
JT9835-3CS 4498.49 USD InStock
JT9835-3 CAJT9835-2 JT9835-2 CAJT9835-3
Acetonitrile with 0.1% (v/v) trifluoroacetic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
Acetonitrile with 0.1% (v/v) Trifluoroacetic acid
For use in liquid chromatography and mass spectrometry. Clear, colorless liquid.


Storage Temperature: Ambient
EINECS: 200-929-3
UN: 1993
ADR: 3,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Trifluoroacetic Acid 0.095 - 0.105 %
Gradient Elution Test (a.u.) - 254 nm ≤0.01
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤200 ppm
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Trace Impurities - * Calcium (Ca) ≤50.0 ppb
Trace Impurities - Iron (Fe) ≤50.0 ppb
Trace Impurities - Magnesium (Mg) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤50 ppb
Trace Impurities - * Sodium (Na) ≤50.0 ppb
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty