Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium 1-pyrrolidinecarbodithioate ≥98.0%, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Pyrrolidinedithiocarbamate ammonium salt1-Pyrrolidinecarbodithioic acid, ammonium saltPyrrolidine-1-Dithiocarboxylic Acid Ammonium SaltAPCD

Management of Change (MOC) category = N

BAKRB337-02 BAKRB337-02
JTB337-2EA 300 USD InStock
CAJTB337-2 JTB337-2
Ammonium 1-pyrrolidinecarbodithioate ≥98.0%, BAKER ANALYZED®, J.T.Baker®
Ammonium 1-pyrrolidinecarbodithioate
Lot analysis on label. For atomic absorption spectroscopy.


Formula: C₅H₁₂N₂S₂
MW: 146.3 · 18.0 g/mol
Melting Pt: ≥146 °C
MDL Number: MFCD00012720
CAS Number: 5108-96-3
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₅H₁₂N₂S₂) ≥98.0 %
Insoluble Matter ≤0.10 %
Melting Point ≥146 °C
Suitability as Chelating Agent Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty