Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95.0-98.0% (by acidimetry) NF, Multi-Compendial, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = R

BAKRCH20-03 BAKRCH20-02
JTCH20-02EA 220.43 USD InStock
JTCH20-02 JTCH20-03
Sulfuric acid 95.0-98.0% (by acidimetry) NF, Multi-Compendial, J.T.Baker®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: ∼335 °C (1013 hPa)
Melting Pt: 3 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Passes test Bulk pharmaceutical chemical
≤ 0.005 % NF - Identification
≤ 0.005 % NF - Residue on Ignition
≤ 1 ppm NF - Chloride (Cl)
Passes Test NF - Arsenic (As)
95.0 - 98.0 % NF - Reducing Substances
Passes Test NF - Assay (as H₂SO₄) (by acidimetry)
Passes Test EP/BP - Identification A
Passes Test EP/BP - Identification B
≤ 50 ppm EP/BP - Appearance of Solution
Passes Test EP/BP - Chloride (Cl)
≤ 25 ppm EP/BP - Nitrate (NO₃)
95.0 - 100.5 % EP/BP - Iron (Fe)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty