Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bromothymol blue sodium salt in aqueous solution, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonateSodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulfonateSodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)-6-isopropyl-3-methylphenolateBromothymol blue water soluble

Management of Change (MOC) category = N

BAKRD472-07 BAKRD472-07
JTD472-7CS 4529.73 USD InStock
CAJTD472-7 JTD472-7
Bromothymol blue sodium salt in aqueous solution, J.T.Baker®
Bromothymol blue sodium salt
Passes alkali consumption test.


Formula: C₂₇H₂₇Br₂NaO₅S
MW: 646.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00077263
CAS Number: 34722-90-2
EINECS: 252-169-7
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Visual Transition Intervals - pH…7.6 (Blue) Passes Test
Visual Transition Intervals - pH…6.7 (Green) Passes Test
Visual Transition Intervals - pH…6.0 (Yellow) Passes Test
Alkali Consumption Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty