Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Butoxyethoxy)ethyl acetate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: Diethylene glycol monobutyl ether acetateDiethyleneglycolmonobutyletheracetate

Management of Change (MOC) category = N

BAKRD660-09 BAKRD660-09
JTD660-9CS 1874.89 USD InStock
CAJTD660-9 JTD660-9
2-(2-Butoxyethoxy)ethyl acetate, J.T.Baker®
2-(2-Butoxyethoxy)ethyl acetate


Formula: C₁₀H₂₀O₄
MW: 204.27 g/mol
Density: 0.99 Kg/L
Flash Pt: 116.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00009458
CAS Number: 124-17-4
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty