Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dextran, M.W. ~ 70,000, low fraction, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

BAKRG200-05 BAKRG200-05
JTG200-5CS 2188.37 USD InStock
CAJTG200-5 JTG200-5
Dextran, M.W. ~ 70,000, low fraction, J.T.Baker®
Dextran
Average molecular weight is 60,000 to 90,000.


Formula: (C₆H₁₀O₅)n
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Appearance Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty