Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethylenetriamine ≥98%, BAKER ANALYZED®, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Diethylenetriamine2,2'-Iminodi(ethylamine)DIEN

Management of Change (MOC) category = N

BAKRH768-07 BAKRH768-07
JTH768-7CS 1404.76 USD InStock
JTH768-7 CAJTH768-7
Diethylenetriamine ≥98%, BAKER ANALYZED®, J.T.Baker®
Diethylenetriamine


Formula: C₄H₁₃N₃
MW: 103.17 g/mol
Boiling Pt: 207 °C (1013 hPa)
Melting Pt: –39 °C
Density: 0.9523 g/cm³ (20 °C)
Flash Pt: 102.00 °C
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8,II
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay ≥98 %
Refractive Index, n²ºD 1.4815 - 1.4845
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty