Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98%, Laboratory Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
Synonyms: Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acidTitriplex® VIIdranal® VIEthylenebis(oxyethylenenitrilo)tetraacetic acidEthylenebis(oxyethylenenitrilo)tetra(acetic acid)Kestranal® FE PAEthylene Glycol Bis(2-aminoethyl Ether)-N,N,N,N-tetraacetic acid

Management of Change (MOC) category = N

BAKRL657-06 BAKRL657-05 BAKRL657-06 BAKRL657-05
JTL657-5CS 7162.07 USD InStock
CAJTL657-6 CAJTL657-5 JTL657-5 JTL657-6
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98%, Laboratory Reagent, J.T.Baker®
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)


Formula: C₁₄H₂₄N₂O₁₀
MW: 380.35 g/mol
Melting Pt: 240…244 °C
Storage Temperature: Ambient
MDL Number: MFCD00004291
CAS Number: 67-42-5
EINECS: 200-651-2
Merck Index: 13,03560
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (C₁₄H₂₄N₂O₁₀) ≥98 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty