Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA magnesium disodium salt hydrate, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Ethylenedinitrilotetra-acetic acid magensium disodium saltKestranal® 2S MgEthylenediaminetetra-acetic acid magensium disodium saltEDTA, Magnesium Derivative, Disodium SaltMagnesium disodium ethylenediaminetetraacetateEdetic acid magensium disodium salt

Management of Change (MOC) category = N

BAKRL704-05 BAKRL704-05
JTL704-5CS 9192.22 USD InStock
JTL704-5 CAJTL704-5
EDTA magnesium disodium salt hydrate, J.T.Baker®
EDTA magnesium disodium salt
EDTA, Magnesium Derivative, Disodium Salt


Formula: C₁₀H₁₂MgN₂Na₂O₈·4H₂O
MW: 358.5 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD03613601
CAS Number: 14402-88-1
EINECS: 238-372-3
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Test for EDTA/Mg Ratio Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty