BAKRM191-05
BAKRM191-07
BAKRM191-04
BAKRM191-09
BAKRM191-01
BAKRM191-07
BAKRM191-01
BAKRM191-04
BAKRM191-09
BAKRM191-05
JTM191-09EA
3595.77
USD
InStock
CAJTM191-09
CAJTM191-07
JTM191-09
CAJTM191-01
JTM191-04
JTM191-01
CAJTM191-05
JTM191-07
JTM191-05
CAJTM191-04
Viral inactivation solution, Biotech Reagent, J.T.Baker®
Viral inactivation solution
J.T.Baker® Viral Inactivation Solution Biotech Reagent is a highly effective, ready-to-use product, specifically developed for detergent-based viral inactivation in biopharmaceutical operations and plasma derived therapy processes. J.T.Baker® Viral Inactivation Solution is formulated with non-ionic detergent, that is readily biodegradable and alkylphenol ethoxylate free, addressing environmental concerns associated with REACH. This innovative solution is optimized to have low protein reactive impurities, low viscosity and compatibility with typical downstream processes. J.T.Baker® Viral Inactivation Solution Biotech Reagent is cGMP and available in a variety of sizes to support development to large scale commercial batches. Detail description provided in technical literature.
- High purity, tested for bioburden and impurity
- Effective inactivation of virus
- Easily cleared in downstream processing, detectable to 1ppm
- Readily biodegradable
- 0.1 µm filtered solution
Proprietary Formulation. Not hazardous according to regulation 29 CFR 1910.1200 (OSHA HCS)