Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Florisil® 60-100 mesh, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: TalcFlorisilSilicic acid magnesium saltMagnesium silicate, activated

Management of Change (MOC) category = N

BAKRM368-08 BAKRM368-08 BAKRM368-07 BAKRM368-07
JTM368-8CS 3904.2 USD InStock
JTM368-8 CAJTM368-7 CAJTM368-8 JTM368-7
Florisil® 60-100 mesh, J.T.Baker®
Magnesium silicate
Selective adsorbent for gas chromatography. Contains activated magnesium silicate.


Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 Kg/L
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Mesh - On U.S. No. 60 Sieve ≤10 %
Mesh - Thru U.S. No. 100 Sieve ≤10 %
Appearance (fine, white crystalline powder). However, due to the high temperature involved in the activation process, some small
charred particles may be visible within the product.
Florisil® - Registered Trademark of Floridin Company

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty