Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium-L(+)-glutamate monohydrate 98.5-101.5% (dry basis) FCC, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: L(+)-α-Glutamic acid sodium salt monohydrate(S)-(+)-2-Aminopentanedioic acid sodium salt monohydrateL(+)-Glutamic acid sodium salt monohydrateSodium L-Glutamate Monohydrate(S)-(+)-Sodium glutamate monohydrateL(+)-2-Aminopentanedioic acid sodium salt monohydrate(S)-(+)-Glutamic acid sodium salt monohydrate(S)-(+)-α-Glutamic acid sodium salt monohydrateL(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrateL(+)-Glutamic acid monosodium salt monohydrate(S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate

Management of Change (MOC) category = RL

J.T. Baker®
BAKRM746-08 BAKRM746-07 BAKRM746-09 M746-08 BAKRM746-09 BAKRM746-07
JTM746-08CS 2043.87 USD InStock
CAJTM746-7 JTM746-08 JTM746-7 CAJTM746-09 CAJTM746-08 JTM746-09
Sodium-L(+)-glutamate monohydrate 98.5-101.5% (dry basis) FCC, J.T.Baker®
Sodium-L(+)-glutamate monohydrate


Formula: C₅H₈NNaO₄·H₂O
MW: 187.13 g/mol
Melting Pt: 225…240 °C
Flash Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00150138
CAS Number: 6106-04-3
EINECS: 240-313-1
Merck Index: 13,06278
Order Now
Specification Test Results
Food GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
FCC - Identification Passes Test
FCC - Assay (dried basis) 98.5 - 101.5 %
FCC - Chloride (Cl) ≤0.2 %
FCC - Lead (Pb) ≤5 mg/kg
FCC - Clarity and Color of Solution Passes Test
FCC - Loss on Drying ≤0.5 %
FCC - Specific Rotation [Å]²ºD (+) 24.5 - 25.5 Degree

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty