Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Hexadecane 90-100%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: CetaneHexadecane

Management of Change (MOC) category = N

BAKRN105-07 BAKRN105-07
JTN105-7CS 3909.91 USD InStock
CAJTN105-7 JTN105-7
n-Hexadecane 90-100%, J.T.Baker®
n-Hexadecane


Formula: H₃C(CH₂)₁₄CH₃
MW: 226.45 g/mol
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.77331 g/cm³ (20 °C)
Flash Pt: 135 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008998
CAS Number: 544-76-3
EINECS: 208-878-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/32/736932.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty