Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-4-methyl-2-pentanone, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: Diacetone alcohol

Management of Change (MOC) category = N

BAKRN679-09 BAKRN679-09
JTN679-9CS 1292.03 USD InStock
CAJTN679-9 JTN679-9
4-Hydroxy-4-methyl-2-pentanone, J.T.Baker®
4-Hydroxy-4-methyl-2-pentanone


Formula: (CH₃)₂C(OH)CH₂COCH₃
MW: 116.16 g/mol
Boiling Pt: 166 °C (1013 hPa)
Melting Pt: –44 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00004471
CAS Number: 123-42-2
EINECS: 204-626-7
UN: 1148
ADR: 3,III
REACH: 01-2119473975-21
Merck Index: 13,02983
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/76/739576.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty