Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kerosene low odor, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Management of Change (MOC) category = N

BAKRP339-00
JTP339-0CS 1316.92 USD InStock
JTP339-0
Kerosene low odor, J.T.Baker®
Kerosene - CAS 8008-20-6


MDL Number: MFCD00135561
CAS Number: 8008-20-6
UN: 1223
ADR: 3,III
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Odor (Mild) Passes Test
Identification (by IR) Passes Test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty