Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methylbutane ≥99.0%, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger
Synonyms: iso-Pentane

Management of Change (MOC) category = N

BAKRQ223-07 BAKRQ223-07 BAKRQ223-08 BAKRQ223-08
JTQ223-8CS 1494.48 USD InStock
JTQ223-8 JTQ223-7 CAJTQ223-8 CAJTQ223-7
2-Methylbutane ≥99.0%, J.T.Baker®
2-Methylbutane


Formula: CH₃CH₂CH(CH₃)₂
MW: 72.15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://us-legacy.vwr.com/stibo/web/std.lang.all/04/68/740468.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Store at or below 68°F (20°C).
Assay ≥99.0 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty