Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Paraffin Oil, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Management of Change (MOC) category = N

BAKRS894-07 BAKRS894-00 BAKRS894-00 BAKRS894-07
JTS894-7CS 948.58 USD InStock
JTS894-7 CAJTS894-0 JTS894-0 CAJTS894-7
Paraffin Oil, J.T.Baker®
Paraffin Oil


Boiling Pt: 300…500 °C (1013 hPa)
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 215 °C (closed cup)
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Identification (by IR) Passes Test
Product Information (not specifications): Saybolt Viscosity @ 100
degrees F (sec).... 345 - 355

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty