Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

Management of Change (MOC) category = N

T254-01 BAKRT254-03 BAKRT254-03 BAKRT254-01
JTT254-1EA 159.9 USD InStock
CAJTT254-3 CAJTT254-1 JTT254-1 JTT254-3
Phenol red, BAKER ANALYZED® ACS indicator, J.T.Baker®
Phenol red


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
Merck Index: 12,07397
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Clarity of Solution Passes Test
Visual Transition Interval - pH…6.8 (Yellow) Passes Test
Visual Transition Interval - pH…8.2 (Red) Passes Test
Meets Reagent Specifications for testing USP/NF monographs

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty